Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010421.4 | NP_034551.2 | 3 - 4 | 473 | 78 | |
GenBank mRNA | AK075895.1 | - | 3 - 4 | 449 | 78 | |
AK075911.1 | - | 3 - 4 | 474 | 78 | ||
AK080777.1 | - | 3 - 4 | 446 | 78 | ||
AK144168.1 | - | 3 - 4 | 601 | 78 | ||
AK150444.1 | - | 3 - 4 | 444 | 78 | ||
AK151960.1 | - | 3 - 4 | 448 | 78 | ||
AK159091.1 | - | 3 - 4 | 465 | 78 | ||
AK159814.1 | - | 3 - 4 | 459 | 78 | ||
AK159890.1 | - | 3 - 4 | 446 | 78 | ||
AK160112.1 | - | 3 - 4 | 433 | 78 | ||
AK167013.1 | - | 3 - 4 | 459 | 78 | ||
AK168186.1 | - | 3 - 4 | 473 | 78 | ||
AK170903.1 | - | 3 - 4 | 459 | 78 | ||
BC010755.1 | - | 3 - 4 | 442 | 4225233 | 78 | |
U07631.1 | - | 3 - 4 | 465 | 78 | ||
X64331.1 | - | 3 - 4 | 416 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |