Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rph3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB112924.1 | - | 7 - 8 | 681 | 78 | |
AK139040.1 | - | 6 - 7 | 78 | |||
BC042585.1 | - | 7 - 8 | 848 | 5364086 | 78 | |
BC050883.1 | - | 7 - 8 | 815 | 5698459 | 78 | |
D29965.1 | - | 7 - 8 | 658 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |