Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013867.2 | NP_038895.1 | 2 - 3 | 715 | 91 | |
GenBank mRNA | AF179566.1 | - | 2 - 3 | 663 | 91 | |
AK143894.1 | - | 2 - 3 | 503 | 91 | ||
AK158241.1 | - | 2 - 3 | 717 | 91 | ||
AK167578.1 | - | 2 - 3 | 534 | 91 | ||
BC023930.1 | - | 2 - 3 | 704 | 5324365 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |