Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013900.2 | NP_038928.1 | 1 - 2 | 167 | 70 | |
GenBank mRNA | AB024336.1 | - | 1 - 2 | 167 | 70 | |
AK036444.1 | - | 1 - 2 | 102 | 70 | ||
BC040347.1 | - | 1 - 2 | 90 | 3672916 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |