Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025285.2 | NP_079561.1 | 3 - 4 | 374 | 126 | |
GenBank mRNA | AK003659.1 | - | 3 - 4 | 374 | 126 | |
AK013020.1 | - | 3 - 4 | 375 | 126 | ||
AK149212.1 | - | 3 - 4 | 399 | 126 | ||
BC026538.1 | - | 3 - 4 | 336 | 4981703 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |