Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159394.1 | NP_001152866.1 | 2 - 3 | 508 | 56 | |
NM_001159395.1 | NP_001152867.1 | 3 - 4 | 390 | 56 | ||
NM_030612.3 | NP_085115.1 | 4 - 5 | 738 | 56 | ||
GenBank mRNA | AB020974.1 | - | 4 - 5 | 950 | 56 | |
AB026551.2 | - | 2 - 3 | 508 | 56 | ||
AB047549.2 | - | 3 - 4 | 403 | 56 | ||
AK082908.1 | - | 2 - 3 | 507 | 56 | ||
AK141318.1 | - | 2 - 3 | 507 | 56 | ||
AK149946.1 | - | 3 - 4 | 386 | 56 | ||
AK150519.1 | - | 2 - 3 | 507 | 56 | ||
AK152016.1 | - | 2 - 3 | 509 | 56 | ||
AK156826.1 | - | 2 - 3 | 509 | 56 | ||
AK159171.1 | - | 2 - 3 | 509 | 56 | ||
AK159459.1 | - | 2 - 3 | 509 | 56 | ||
AK159917.1 | - | 2 - 3 | 509 | 56 | ||
AK160065.1 | - | 2 - 3 | 507 | 56 | ||
BC058188.1 | - | 2 - 3 | 493 | 5341736 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |