Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159394.1 | NP_001152866.1 | 5 - 6 | 1453 | 97 | |
NM_001159395.1 | NP_001152867.1 | 6 - 7 | 1335 | 97 | ||
NM_030612.3 | NP_085115.1 | 7 - 8 | 1683 | 97 | ||
GenBank mRNA | AB020974.1 | - | 7 - 8 | 1895 | 97 | |
AB026551.2 | - | 5 - 6 | 1453 | 97 | ||
AB047549.2 | - | 6 - 7 | 1348 | 97 | ||
AK082908.1 | - | 5 - 6 | 1452 | 97 | ||
AK136509.1 | - | 2 - 3 | 922 | 97 | ||
AK141318.1 | - | 5 - 6 | 1454 | 97 | ||
AK149946.1 | - | 6 - 7 | 1331 | 97 | ||
AK150519.1 | - | 5 - 6 | 1452 | 97 | ||
AK152016.1 | - | 5 - 6 | 1454 | 97 | ||
AK156826.1 | - | 5 - 6 | 1453 | 97 | ||
AK159171.1 | - | 5 - 6 | 1454 | 97 | ||
AK159459.1 | - | 5 - 6 | 1454 | 97 | ||
AK159917.1 | - | 5 - 6 | 1454 | 97 | ||
AK160065.1 | - | 5 - 6 | 1452 | 97 | ||
BC058188.1 | - | 5 - 6 | 1438 | 5341736 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |