Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138583.2 | NP_613049.2 | 3 - 4 | 222 | 96 | |
GenBank mRNA | AK089239.1 | - | 3 - 4 | 204 | 96 | |
AK133588.1 | - | 3 - 4 | 205 | 96 | ||
AK168595.1 | - | 2 - 3 | 201 | 96 | ||
BC005445.1 | - | 3 - 4 | 283 | 3496749 | 96 | |
X74504.1 | - | 2 - 3 | 195 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |