Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P478881 |
BC026374.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P478881 |
NM_146213.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P478881 |
AK037191.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P478881 |
BC026374.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P478881 |
NM_146213.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P478881 |
AK037191.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AF061017.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK054064.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK144673.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK146693.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK162298.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK167462.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK167560.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK168064.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK168508.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK168563.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
AK169208.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
BC006749.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
500013 |
NM_009466.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
877131 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
92920 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
80715 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
899716 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
631689 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
131810 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
462319 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
448355 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240475 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
761450 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
764515 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
647552 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
55057 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
71594 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P122080 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P107029 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P478881 |
BC026374.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P478881 |
NM_146213.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P478881 |
AK037191.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P478881 |
BC026374.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P478881 |
NM_146213.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P478881 |
AK037191.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131802 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131820 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131816 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131813 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131782 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131795 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131805 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131806 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131798 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131784 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131790 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131781 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131812 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131811 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131819 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131797 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131801 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131807 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131800 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131787 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131792 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131788 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131814 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26131808 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P122080 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P107029 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P478881 |
BC026374.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P478881 |
NM_146213.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P478881 |
AK037191.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P478881 |
BC026374.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P478881 |
NM_146213.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P478881 |
AK037191.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132744 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P122080 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P107029 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06836188 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06836183 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06836185 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06836177 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06836180 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06836181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131885 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131870 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131895 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131937 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131964 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131892 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131906 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131951 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131893 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131910 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131948 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26131941 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131775 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131957 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07216706 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131816 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06809316 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131777 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131794 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131948 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131945 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131773 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07198235 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07116199 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06800209 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131921 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131941 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131787 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06908381 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131802 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131936 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131811 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131826 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07198238 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07055337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131793 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131820 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131954 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131822 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131924 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131799 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131946 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131824 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131781 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131783 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131835 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131808 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131930 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131782 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131927 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07076545 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07174795 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131956 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131934 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131829 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131933 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131830 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131955 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131922 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131817 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131837 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06836177 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06813565 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131940 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131937 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07436896 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131795 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06908382 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06723517 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131788 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131805 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07400350 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07314410 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131778 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26131951 |
- |
- |