Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022378.3 | NP_071773.2 | 1 - 2 | 391 | 80 | |
GenBank mRNA | BC111908.1 | - | 1 - 2 | 102 | 40045942 | 80 |
U04198.1 | - | 1 - 2 | 298 | 80 | ||
U90538.1 | - | 1 - 2 | 338 | 80 | ||
X92592.1 | - | 1 - 2 | 279 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |