Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024200.4 | NP_077162.2 | 1 - 2 | 210 | 62 | |
GenBank mRNA | AK018181.1 | - | 1 - 2 | 207 | 62 | |
AK141611.1 | - | 1 - 2 | 211 | 62 | ||
AK220169.1 | - | 1 - 2 | 198 | 62 | ||
BC056641.1 | - | 1 - 2 | 169 | 5097345 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |