Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174850.3 | NP_777275.2 | 12 - 13 | 2807 | 112 | |
GenBank mRNA | AB182579.1 | - | 12 - 13 | 2658 | 112 | |
AK044547.1 | - | 12 - 13 | 2659 | 112 | ||
AK151086.1 | - | 12 - 13 | 2806 | 112 | ||
AK164043.1 | - | 12 - 13 | 2778 | 112 | ||
AK165558.1 | - | 12 - 13 | 2657 | 112 | ||
AK220308.1 | - | 12 - 13 | 2584 | 112 | ||
BC043300.1 | - | 12 - 13 | 2621 | 5025620 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |