Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175111.3 | NP_780320.2 | 1 - 2 | 152 | 97 | |
GenBank mRNA | AA879889.1 | - | 1 - 2 | 154 | 97 | |
AK017731.1 | - | 1 - 2 | 118 | 97 | ||
AK028824.1 | - | 1 - 2 | 133 | 97 | ||
AK049404.1 | - | 1 - 2 | 130 | 97 | ||
AK076189.1 | - | 1 - 2 | 119 | 97 | ||
AK089941.1 | - | 1 - 2 | 167 | 97 | ||
AK138906.1 | - | 1 - 2 | 132 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |