Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289545.1 | NP_001276474.1 | 9 - 10 | 1509 | 75 | |
NM_175183.5 | NP_780392.1 | 10 - 11 | 1688 | 75 | ||
GenBank mRNA | AK046849.1 | - | 10 - 11 | 2096 | 75 | |
BC027136.1 | - | 6 - 7 | 1986 | 75 | ||
BC088996.1 | - | 8 - 9 | 1283 | 75 | ||
BC094547.1 | - | 9 - 10 | 1443 | 75 | ||
BC137599.1 | - | 9 - 10 | 1430 | 8860618 | 75 | |
BC137600.1 | - | 9 - 10 | 1457 | 8860619 | 75 | |
BC144756.1 | - | 9 - 10 | 1437 | 9053304 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |