Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252494.1 | NP_001239423.1 | 3 - 4 | 399 | 119 | |
NM_001252496.1 | NP_001239425.1 | 3 - 4 | 399 | 119 | ||
NM_175258.4 | NP_780467.2 | 3 - 4 | 399 | 119 | ||
GenBank mRNA | AK045540.1 | - | 3 - 4 | 341 | 119 | |
AK049228.1 | - | 3 - 4 | 355 | 119 | ||
AK167055.1 | - | 3 - 4 | 400 | 119 | ||
BC141182.1 | - | 3 - 4 | 298 | 9055794 | 119 | |
BC145503.1 | - | 3 - 4 | 298 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |