Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052340.1 | - | 11 - 12 | 1448 | 66 | |
AK173233.1 | - | 8 - 9 | 837 | 66 | ||
BC059083.1 | - | 11 - 12 | 1448 | 6827289 | 66 | |
BC062917.1 | - | 11 - 12 | 1454 | 6831835 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |