Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159672.1 | NP_001153144.1 | 10 - 11 | 1116 | 58 | |
NM_175317.3 | NP_780526.2 | 10 - 11 | 1145 | 58 | ||
GenBank mRNA | AK028676.1 | - | 10 - 11 | 1118 | 58 | |
AK031647.1 | - | 10 - 11 | 1102 | 58 | ||
AK049698.1 | - | 10 - 11 | 1104 | 58 | ||
BC031852.1 | - | 4 - 5 | 391 | 58 | ||
BC045616.1 | - | 10 - 11 | 1099 | 4983234 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |