Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175324.3 | NP_780533.2 | 6 - 7 | 897 | 76 | |
GenBank mRNA | AK077531.1 | - | 6 - 7 | 898 | 76 | |
AK128998.1 | - | 6 - 7 | 876 | 76 | ||
AK139918.1 | - | 6 - 7 | 841 | 76 | ||
AK145561.1 | - | 6 - 7 | 886 | 76 | ||
BC045199.1 | - | 6 - 7 | 861 | 5133197 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |