Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175341.4 | NP_780550.1 | 3 - 4 | 1023 | 87 | |
NM_207515.2 | NP_997398.1 | 3 - 4 | 1023 | 87 | ||
GenBank mRNA | AK028797.1 | - | 3 - 4 | 988 | 87 | |
AK085709.1 | - | 3 - 4 | 986 | 87 | ||
AK151913.1 | - | 3 - 4 | 987 | 87 | ||
AK151926.1 | - | 3 - 4 | 987 | 87 | ||
AK151957.1 | - | 3 - 4 | 987 | 87 | ||
AK153261.1 | - | 3 - 4 | 877 | 87 | ||
AK159213.1 | - | 3 - 4 | 981 | 87 | ||
AK164372.1 | - | 3 - 4 | 986 | 87 | ||
AK167479.1 | - | 3 - 4 | 853 | 87 | ||
AK167694.1 | - | 4 - 5 | 789 | 87 | ||
AK220470.1 | - | 997 | 87 | |||
BC020406.1 | - | 2 - 3 | 421 | 87 | ||
BC023895.1 | - | 3 - 4 | 975 | 87 | ||
BC026711.1 | - | 2 - 3 | 388 | 87 | ||
BC075665.1 | - | 2 - 3 | 626 | 30605146 | 87 | |
BC079898.1 | - | 3 - 4 | 972 | 87 | ||
GQ906534.1 | - | 2 - 3 | 341 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |