Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175360.2 | NP_780569.1 | 4 - 5 | 551 | 73 | |
GenBank mRNA | AK075574.1 | - | 4 - 5 | 551 | 73 | |
AK135228.1 | - | 4 - 5 | 569 | 73 | ||
AK146929.1 | - | 4 - 5 | 629 | 73 | ||
AK164386.1 | - | 4 - 5 | 561 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |