Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK046449.1 | - | 1 | 2149 | 101 | |
AK081215.1 | - | 1 | 824 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |