Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175430.4 | NP_780639.1 | 10 - 11 | 2656 | 60 | |
GenBank mRNA | AK046971.1 | - | 10 - 11 | 2659 | 60 | |
AK054172.1 | - | 11 - 12 | 2170 | 60 | ||
BC072591.1 | - | 12 - 13 | 2385 | 60 | ||
BC085159.1 | - | 12 - 13 | 2416 | 30533905 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |