Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clvs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175448.3 | NP_780657.1 | 1 - 2 | 464 | 63 | |
GenBank mRNA | AK039308.1 | - | 1 - 2 | 465 | 63 | |
AK053797.1 | - | 1 - 2 | 487 | 63 | ||
AK139409.1 | - | 1 - 2 | 539 | 64 | ||
AK139868.1 | - | 1 - 2 | 517 | 63 | ||
AK163214.1 | - | 1 - 2 | 485 | 63 | ||
AK169009.1 | - | 1 - 2 | 634 | 63 | ||
BC058539.1 | - | 1 - 2 | 471 | 5690124 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |