Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175482.3 | NP_780691.2 | 17 - 18 | 2281 | 78 | |
GenBank mRNA | AK028335.1 | - | 3 - 4 | 476 | 78 | |
AK036673.1 | - | 7 - 8 | 1054 | 78 | ||
AK052442.1 | - | 9 - 10 | 1314 | 78 | ||
AK129380.1 | - | 7 - 8 | 1014 | 78 | ||
BC066033.1 | - | 17 - 18 | 2177 | 6835629 | 78 | |
BC088733.1 | - | 17 - 18 | 2203 | 30459522 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |