Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175501.2 | NP_780710.2 | 11 - 12 | 1976 | 74 | |
GenBank mRNA | AJ537452.1 | - | 11 - 12 | 1735 | 74 | |
AK028459.1 | - | 2 - 3 | 233 | 74 | ||
AK048612.1 | - | 11 - 12 | 1939 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |