Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P409457 |
BC089351.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P409457 |
AK046751.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P409457 |
NM_176831.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P409457 |
BC089351.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P409457 |
AK046751.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P409457 |
NM_176831.4 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
952041 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
649407 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
13614 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
413246 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
279434 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
520337 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
662283 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
152209 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
296348 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435623 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
495473 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
501336 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
291141 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
483962 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
481444 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
489507 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
167071 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
241240 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505492 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
27140 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
706751 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
320157 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
573590 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
85408 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506975 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
524011 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
459837 |
BC080690.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
459837 |
NM_001013405.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
381576 |
AF039213.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
381576 |
AK083174.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
381576 |
NM_008432.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497054 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384793 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777083 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
50948 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
69292 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
175374 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
11334 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P158411 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P176156 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P409457 |
BC089351.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P409457 |
AK046751.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P409457 |
NM_176831.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P409457 |
BC089351.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P409457 |
AK046751.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P409457 |
NM_176831.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P135022 |
BC004779.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P135022 |
BC052928.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P135022 |
BC089351.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P135022 |
AK039846.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P135022 |
AK046751.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541860 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541850 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541847 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541842 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541857 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541852 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541844 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541854 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541851 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541840 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541845 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26541855 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06801596 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P158411 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P176156 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01941660 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06801569 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06801633 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P409457 |
BC089351.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P409457 |
AK046751.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P409457 |
NM_176831.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P409457 |
BC089351.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P409457 |
AK046751.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P409457 |
NM_176831.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P135022 |
BC004779.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P135022 |
BC052928.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P135022 |
BC089351.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P135022 |
AK039846.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P135022 |
AK046751.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105485 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P107604 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P158411 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P176156 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06801569 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07466820 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07979161 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07555411 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07979160 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07555410 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07833470 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07555414 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07979159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541840 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541704 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541850 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541824 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541845 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541752 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541800 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541720 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541713 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26541772 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07508485 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541840 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541838 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07545709 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541729 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541839 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541834 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07707878 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07508460 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541744 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541811 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541746 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541827 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541777 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07979159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07749417 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541739 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07880756 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07651910 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07545714 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541844 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07545728 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07880758 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541841 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541807 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07545708 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541772 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541780 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541726 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07555410 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541733 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07545698 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541815 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541774 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541816 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07545717 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541824 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541794 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541731 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07545724 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541742 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541831 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541775 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07880760 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541855 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541842 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541845 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541849 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541819 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541745 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541730 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541822 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541768 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541836 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541737 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541857 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541805 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541850 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541748 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541798 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541847 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26541860 |
- |
- |