Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176838.2 | NP_789808.1 | 12 - 13 | 1921 | 56 | |
GenBank mRNA | AK088755.1 | - | 12 - 13 | 1791 | 56 | |
AK168488.1 | - | 11 - 12 | 1154 | 56 | ||
BC031444.1 | - | 12 - 13 | 1921 | 4222237 | 56 | |
BC096581.1 | - | 12 - 13 | 1758 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |