Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176840.3 | NP_789810.2 | 1 - 2 | 932 | 151 | |
GenBank mRNA | BC035278.1 | - | 1 - 2 | 932 | 5043236 | 151 |
BC099400.1 | - | 1 - 2 | 847 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |