Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177333.3 | NP_796307.2 | 2 - 3 | 239 | 68 | |
GenBank mRNA | AK088333.1 | - | 2 - 3 | 233 | 68 | |
AK089979.1 | - | 2 - 3 | 250 | 68 | ||
AK131141.1 | - | 2 - 3 | 223 | 68 | ||
AK141537.1 | - | 2 - 3 | 239 | 68 | ||
BC031743.1 | - | 2 - 3 | 227 | 4187527 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |