Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gldn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177350.5 | NP_796324.1 | 8 - 9 | 1055 | 61 | |
GenBank mRNA | AF548022.1 | - | 8 - 9 | 711 | 61 | |
AK031523.1 | - | 8 - 9 | 1056 | 61 | ||
BC125025.1 | - | 8 - 9 | 1035 | 40058551 | 61 | |
BC125026.1 | - | 8 - 9 | 1035 | 40058552 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |