Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177363.3 | NP_796337.1 | 3 - 4 | 445 | 74 | |
GenBank mRNA | AK036904.1 | - | 3 - 4 | 446 | 74 | |
AK089206.1 | - | 74 | ||||
AK155804.1 | - | 3 - 4 | 453 | 74 | ||
AK170277.1 | - | 2 - 3 | 376 | 74 | ||
DQ973493.1 | - | 3 - 4 | 397 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |