Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27561411 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27561412 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27561413 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2092760 |
AK156451.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2092760 |
BC062155.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2092760 |
BC096693.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2092760 |
NM_177370.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2092760 |
AK053223.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363815 |
AK002395.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363815 |
AK005237.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
363815 |
NM_008618.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
69215 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809227 |
AK017346.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809227 |
AK046748.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809227 |
AK077921.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809227 |
BC099971.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
809227 |
NM_183186.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
591976 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P142220 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2092760 |
AK156451.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2092760 |
BC062155.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2092760 |
BC096693.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2092760 |
NM_177370.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2092760 |
AK053223.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561423 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561418 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561442 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561426 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561430 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561421 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561453 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561424 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561431 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561429 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561419 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561432 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561416 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561436 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561438 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561428 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561412 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561440 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27561450 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P142220 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P114563 |
AK156451.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P114563 |
BC062155.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P114563 |
BC096693.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P114563 |
NM_177370.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P114563 |
AK053223.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113223 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P142220 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09095171 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09095193 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27561411 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27561413 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27561412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561409 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561457 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27561455 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561426 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561450 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561455 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561438 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561461 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561441 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561418 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561409 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561466 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561452 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561459 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561431 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561444 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561468 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09577027 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561442 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09440424 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561464 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561428 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561433 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561419 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561440 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561416 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561423 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561454 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561421 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561436 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561448 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09577020 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09434263 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27561424 |
- |
- |