Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177595.4 | NP_808263.2 | 2 - 3 | 432 | 76 | |
GenBank mRNA | AK034675.1 | - | 2 - 3 | 410 | 76 | |
BC137728.1 | - | 2 - 3 | 367 | 8860747 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |