Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159883.1 | NP_001153355.1 | 5 - 6 | 558 | 71 | |
NM_001159884.1 | NP_001153356.1 | 5 - 6 | 506 | 71 | ||
NM_001159885.1 | NP_001153357.1 | 4 - 5 | 541 | 71 | ||
NM_020266.2 | NP_064662.2 | 4 - 5 | 504 | 71 | ||
NM_178055.4 | NP_835156.1 | 5 - 6 | 558 | 71 | ||
GenBank mRNA | AK076061.1 | - | 5 - 6 | 494 | 71 | |
AK156083.1 | - | 5 - 6 | 527 | 71 | ||
AK171494.1 | - | 5 - 6 | 504 | 71 | ||
BC011090.1 | - | 5 - 6 | 460 | 3968468 | 71 | |
BC085162.1 | - | 2 - 3 | 2667 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |