Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290633.1 | NP_001277562.1 | 2 - 3 | 513 | 75 | |
NM_178256.4 | NP_839987.2 | 2 - 3 | 513 | 75 | ||
GenBank mRNA | AK147418.1 | - | 2 - 3 | 164 | 75 | |
AY263368.1 | - | 2 - 3 | 458 | 75 | ||
BC139012.1 | - | 2 - 3 | 389 | 8862034 | 75 | |
BC145500.1 | - | 2 - 3 | 389 | 9054063 | 75 | |
CJ123995.1 | - | 2 - 3 | 516 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |