Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178379.3 | NP_848466.1 | 1 - 2 | 137 | 88 | |
GenBank mRNA | AK010385.1 | - | 1 - 2 | 131 | 88 | |
AK149999.1 | - | 1 - 2 | 127 | 88 | ||
AK154668.1 | - | 1 - 2 | 136 | 88 | ||
AK156865.1 | - | 1 - 2 | 136 | 88 | ||
AK171661.1 | - | 1 - 2 | 127 | 88 | ||
AK172718.1 | - | 1 - 2 | 136 | 88 | ||
AK213695.1 | - | 1 - 2 | 38 | 88 | ||
BC018301.1 | - | 1 - 2 | 89 | 88 | ||
BC038046.1 | - | 1 - 2 | 90 | 4206826 | 88 | |
BY078593.1 | - | 1 - 2 | 137 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |