Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178387.3 | NP_848474.2 | 6 - 7 | 926 | 73 | |
GenBank mRNA | AK006916.1 | - | 5 - 6 | 604 | 73 | |
BC050799.1 | - | 5 - 6 | 828 | 73 | ||
BC120503.1 | - | 3 - 4 | 378 | 73 | ||
BC120505.1 | - | 3 - 4 | 378 | 73 | ||
BC144721.1 | - | 3 - 4 | 378 | 73 | ||
BC144723.1 | - | 3 - 4 | 378 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |