Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178392.4 | NP_848479.1 | 7 - 8 | 915 | 124 | |
GenBank mRNA | AK012317.1 | - | 6 - 7 | 572 | 124 | |
AK138261.1 | - | 7 - 8 | 916 | 124 | ||
AK152833.1 | - | 7 - 8 | 921 | 124 | ||
BC030456.1 | - | 7 - 8 | 897 | 5401615 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |