Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178405.3 | NP_848492.1 | 3 - 4 | 350 | 76 | |
GenBank mRNA | AK052863.1 | - | 3 - 4 | 286 | 76 | |
AK129212.1 | - | 3 - 4 | 263 | 76 | ||
AK137602.1 | - | 3 - 4 | 286 | 76 | ||
AK147331.1 | - | 3 - 4 | 282 | 76 | ||
AK148775.1 | - | 3 - 4 | 282 | 76 | ||
AK161840.1 | - | 3 - 4 | 282 | 76 | ||
AK164509.1 | - | 3 - 4 | 286 | 76 | ||
AK168703.1 | - | 3 - 4 | 282 | 76 | ||
BC036127.1 | - | 3 - 4 | 274 | 4955003 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |