Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178592.3 | NP_848707.1 | 6 - 7 | 540 | 98 | |
GenBank mRNA | BC029114.1 | - | 6 - 7 | 540 | 2648618 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |