Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178639.4 | NP_848754.1 | 8 - 9 | 475 | 65 | |
GenBank mRNA | AF325264.1 | - | 8 - 9 | 493 | 65 | |
AK036285.1 | - | 8 - 9 | 473 | 65 | ||
AK043706.1 | - | 8 - 9 | 474 | 65 | ||
AK082073.1 | - | 8 - 9 | 473 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |