Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178654.4 | NP_848769.2 | 20 - 21 | 3015 | 127 | |
GenBank mRNA | AK049713.1 | - | 20 - 21 | 3002 | 127 | |
AK054281.1 | - | 6 - 7 | 619 | 127 | ||
AK083425.1 | - | 15 - 16 | 1813 | 127 | ||
AK133298.1 | - | 20 - 21 | 3004 | 127 | ||
AK171092.1 | - | 20 - 21 | 2946 | 127 | ||
BC052073.1 | - | 21 - 22 | 2938 | 6400440 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |