Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178654.4 | NP_848769.2 | 3 - 4 | 853 | 78 | |
GenBank mRNA | AK028469.1 | - | 3 - 4 | 840 | 78 | |
AK046248.1 | - | 3 - 4 | 770 | 78 | ||
AK049713.1 | - | 3 - 4 | 840 | 78 | ||
AK077910.1 | - | 3 - 4 | 833 | 78 | ||
AK133298.1 | - | 3 - 4 | 842 | 78 | ||
AK171092.1 | - | 3 - 4 | 784 | 78 | ||
BC052073.1 | - | 4 - 5 | 776 | 6400440 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |