Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178662.3 | NP_848777.1 | 11 - 12 | 1417 | 66 | |
GenBank mRNA | AK028298.1 | - | 11 - 12 | 1424 | 66 | |
AK038483.1 | - | 11 - 12 | 1427 | 66 | ||
AK038990.1 | - | 11 - 12 | 1418 | 66 | ||
AK039411.1 | - | 11 - 12 | 1425 | 66 | ||
AK086723.1 | - | 11 - 12 | 1460 | 66 | ||
AK162959.1 | - | 11 - 12 | 1425 | 66 | ||
AY349150.1 | - | 11 - 12 | 1382 | 66 | ||
BC048903.1 | - | 11 - 12 | 1401 | 6491141 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |