Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178676.4 | NP_848791.2 | 7 - 8 | 906 | 69 | |
GenBank mRNA | AK032057.1 | - | 7 - 8 | 892 | 69 | |
AK046218.1 | - | 7 - 8 | 882 | 69 | ||
AK087561.1 | - | 7 - 8 | 880 | 69 | ||
AK169947.1 | - | 2 - 3 | 316 | 69 | ||
AK170974.1 | - | 2 - 3 | 315 | 69 | ||
AY376710.1 | - | 7 - 8 | 874 | 69 | ||
AY714060.1 | - | 7 - 8 | 877 | 69 | ||
BC079871.1 | - | 7 - 8 | 877 | 6831925 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |