Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145877.2 | NP_001139349.1 | 1 - 2 | 417 | 62 | |
NM_178696.5 | NP_848811.2 | 1 - 2 | 417 | 62 | ||
GenBank mRNA | AK032960.1 | - | 1 - 2 | 395 | 62 | |
AK046586.1 | - | 1 - 2 | 380 | 62 | ||
AK079532.1 | - | 1 - 2 | 381 | 62 | ||
AK083273.1 | - | 1 - 2 | 387 | 62 | ||
AK134914.1 | - | 1 - 2 | 416 | 62 | ||
AK165159.1 | - | 1 - 2 | 384 | 62 | ||
AK166011.1 | - | 1 - 2 | 389 | 62 | ||
AK166331.1 | - | 1 - 2 | 378 | 62 | ||
AK168955.1 | - | 1 - 2 | 390 | 62 | ||
AK169886.1 | - | 1 - 2 | 381 | 62 | ||
BC012234.1 | - | 1 - 2 | 196 | 4191504 | 62 | |
BC052771.1 | - | 1 - 2 | 362 | 6310271 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |