Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178931.2 | NP_849262.1 | 1 - 2 | 102 | 63 | |
GenBank mRNA | AF515707.1 | - | 1 - 2 | 71 | 63 | |
AY264405.1 | - | 1 - 2 | 102 | 63 | ||
BC104054.1 | - | 1 - 2 | 102 | 40040309 | 63 | |
BC104055.1 | - | 1 - 2 | 102 | 40040311 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |