Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178931.2 | NP_849262.1 | 3 - 4 | 330 | 99 | |
GenBank mRNA | AF515707.1 | - | 3 - 4 | 299 | 99 | |
AY264405.1 | - | 3 - 4 | 330 | 99 | ||
BC022125.1 | - | 1 - 2 | 61 | 99 | ||
BC104054.1 | - | 3 - 4 | 330 | 40040309 | 99 | |
BC104055.1 | - | 3 - 4 | 330 | 40040311 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |