Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181404.5 | NP_852069.4 | 7 - 8 | 3434 | 150 | |
GenBank mRNA | AK122214.1 | - | 3 - 4 | 1800 | 150 | |
AK147945.1 | - | 4 - 5 | 3021 | 150 | ||
BC050250.1 | - | 2 - 3 | 241 | 151 | ||
BC079563.1 | - | 3 - 4 | 2761 | 30535512 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |