Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300007378.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007378.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007378.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007380.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007380.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007380.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007379.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007379.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007379.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007384.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007384.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007384.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007376.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007376.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007376.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007377.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007377.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007377.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007386.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007386.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007386.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007382.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007382.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007382.mm.1 |
TC0300000925.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007383.mm.1 |
NM_181409.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007383.mm.1 |
NM_181409.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007383.mm.1 |
TC0300000925.mm.1 |
MTA TC |